Imprimer
Hématoxyline en solution selon Harris (modifié) solution (Harris) stabilisé exempt de mercure
Fournisseur: RAL (REACTIFS RAL SA)
Synonyms: Hematoxylin Harris
RALA361075-1000EA 92.1 CHF
RALA361075-1000
Hématoxyline en solution selon Harris (modifié) solution (Harris) stabilisé exempt de mercure
Hématoxyline en solution selon Harris (modifié)
Harris Haematoxylin stabilized is used as a nuclear staining solution in Papanicolaou staining, progressive variation for the trichromic staining of cervical smears. Harris Haematoxylin stabilized solution (mercury-free) colours nuclei more or less dark violet-blue.
IVDLogo