Imprimer
Vertical stand with electrode holder, Orion™
Fournisseur: Thermo Scientific
662-1138EA 217 CHF
ORIO090070
Vertical stand with electrode holder, Orion™
  • Holds one to three electrodes with 12 mm diameter
  • Vertical motion allows electrodes to be moved up and down into beakers or sample containers
  • Can be used with Orion pH, ORP and ISE (ion selective electrode) benchtop or portable meters
Recommended for Orion A, A+, Thermo Scientific™ PerpHect™ and pHuture™ Series meters.