Imprimer

2-Fluoro-4-(pentafluorothio)benzamide 97%

Fournisseur: Apollo Scientific

APOSPC302451-1GEA 419 CHF
APOSPC302451-1G
2-Fluoro-4-(pentafluorothio)benzamide 97%
2-Fluoro-4-(pentafluorothio)benzamide
Formule: C₇H₅F₆NOS
MW: 265,17 g/mol
température de stockage: Température ambiante
MDL Number: MFCD16652504
Numéro CAS: 1240257-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR