Imprimer

4-Fluoro-1-naphtonitrile 97%

Fournisseur: Apollo Scientific
Attention

APOSPC2102L-1GEA 17.6 CHF
APOSPC2102L-1G
4-Fluoro-1-naphtonitrile 97%
4-Fluoro-1-naphtonitrile
Formule: C₁₁H₆FN
MW: 171,18 g/mol
Melting Pt: 89…91 °C
température de stockage: Température ambiante
MDL Number: MFCD00236656
Numéro CAS: 13916-99-9
ONU: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR