Imprimer

2'-Cloroacetoacetanilida, grade technique

Fournisseur: Apollo Scientific

APOSOR32301-1GEA 1660 CHF
APOSOR32301-1G
2'-Cloroacetoacetanilida, grade technique
2'-Cloroacetoacetanilida
Formule: C₁₀H₁₀ClNO₂
MW: 211,65 g/mol
température de stockage: Température ambiante
MDL Number: MFCD00018224
Numéro CAS: 93-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR