Imprimer

2-Amino-4-chlorobenzamide

Fournisseur: Apollo Scientific

APOSOR307693-1GEA 78.6 CHF
APOSOR307693-1G
2-Amino-4-chlorobenzamide
2-Amino-4-chlorobenzamide
Formule: C₇H₇ClN₂O
MW: 170,6000 g/mol
température de stockage: Température ambiante
MDL Number: MFCD09042792
Numéro CAS: 5900-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR