2-(4-Chlorophenoxy)nicotinic acid
Fournisseur: Apollo Scientific
APOSOR21701-1GEA
463
CHF
APOSOR21701-1G
2-(4-Chlorophenoxy)nicotinic acid
2-(4-Chlorophenoxy)nicotinic acid
Formule:
C₁₂H₈ClNO₃ MW: 249,64 g/mol température de stockage: Température ambiante |
MDL Number:
MFCD00052116 Numéro CAS: 51362-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...