Imprimer

N-Acetyl-D-methionine

APOSBIA1775-10GEA 0 CHF
APOSBIA1775-10G APOSBIA1775-25G
N-Acetyl-D-methionine
N-Acetyl-D-methionine
Formule: C₇H₁₃NO₃S
MW: 191,2 g/mol
température de stockage: Réfrigérateur
MDL Number: MFCD00069962
Numéro CAS: 1509-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR