Imprimer

3,6-Dichloropyridazine

Fournisseur: Molekula

MOLEM12760409EA 0 CHF
MOLEM12760409
3,6-Dichloropyridazine
3,6-Dichloropyridazine
Formule: C₄H₂Cl₂N₂
MW: 148,98 g/mol
Point d'ébullition: 89…91 °C (0,2 mmHg)
Melting Pt: 70 °C
Densité: 1,7 g/cm³ (20 °C)
MDL Number: MFCD00006466
Numéro CAS: 141-30-0
ONU: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR