Imprimer

Spironolactone

EDQMS1200000EA 199 CHF
EDQMS1200000
Spironolactone
Spironolactone
Formule: C₂₄H₃₂O₄S
Point d'ébullition: 597 °C (1013 hPa)
Melting Pt: 196 °C
MDL Number: MFCD00082250
Numéro CAS: 52-01-7
Merck Index: 12,08917

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR