Imprimer

5-Hydroxy-2-hydroxyméthyl-4-pyrone

Fournisseur: Molekula

Synonyms: 5-Hydroxy-2-hydroxyméthyl-γ-pyrone

MOLE31686564-25GEA 0 CHF
MOLE31686564-25G
5-Hydroxy-2-hydroxyméthyl-4-pyrone
5-Hydroxy-2-hydroxyméthyl-4-pyrone
Formule: C₆H₆O₄
MW: 142,11 g/mol
Point d'ébullition: 323 °C (1013 hPa)
Melting Pt: 149…152 °C
Densité: 1,58 g/cm³ (20 °C)
MDL Number: MFCD00006580
Numéro CAS: 501-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR