Imprimer

1-Naphthyl boronic acid

Fournisseur: Molekula

MOLE39657844-1GEA 0 CHF
MOLE39657844-1G
1-Naphthyl boronic acid
1-Naphthyl boronic acid
Formule: C₁₀H₉BO₂
MW: 171,99 g/mol
Melting Pt: 259 °C
MDL Number: MFCD00019722
Numéro CAS: 13922-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR