Imprimer

Ammonium fluorure, BAKER ANALYZED® ACS, J.T.Baker®

Fournisseur: Avantor

Synonyms: Fluorure d'ammonium

0023.0250EA 0 CHF
0023.0250
Ammonium fluorure, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium fluorure
Formule: NH₄F
MW: 37,04 g/mol
Melting Pt: 160 °C
Densité: 1,0088…1.0096 g/cm³ (25 °C)
MDL Number: MFCD00011423
Numéro CAS: 12125-01-8
ONU: 2505
ADR: 6.1,III
Merck Index: 12,00553

Order Now

Résultats des tests de spécification

Meets ACS Specifications
Assay min. 98.0 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 5 ppm
Heavy Metals (as Pb) max. 5 ppm
Insoluble Matter max. 0.005 %
Residue after Ignition max. 0.01 %
Sulfate (SO4) max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR