Imprimer

Chlorodiphénylphosphine

Fournisseur: Molekula

Synonyms: DPPC

MOLEM36302932EA 0 CHF
MOLEM36302932
Chlorodiphénylphosphine
Chlorodiphénylphosphine
Formule: C₁₂H₁₀ClP
MW: 220,64 g/mol
Point d'ébullition: 320 °C (1013 hPa)
Melting Pt: 14…16 °C
Densité: 1,2 g/cm³ (20 °C)
Point d'éclair: 138 °C
MDL Number: MFCD00000529
Numéro CAS: 1079-66-9
ONU: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR