α-Bromo-o-iodotoluène
Fournisseur: Molekula
Synonyms:
α-Bromo-2-iodotoluène
MOLE55965489-5GEA
0
CHF
MOLE55965489-5G
α-Bromo-o-iodotoluène
α-Bromo-o-iodotoluène
Formule:
C₇H₆BrI MW: 296,93 g/mol Point d'ébullition: 279 °C (1013 hPa) Melting Pt: 55,5 °C |
MDL Number:
MFCD00236046 Numéro CAS: 40400-13-3 ONU: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...