Imprimer

Thimerosal

Fournisseur: Enzo Life Sciences

Synonyms: Sodium (éthylmercurithio)-2 benzoateMerthiolatSodium mercurothiolate

ALEXALX400013-G005EA 0 CHF
ALEXALX400013-G005
Thimerosal
Thimerosal
Formule: 2-(C₂H₅HgS)C₆H₄CO₂Na
MW: 404,82 g/mol
Melting Pt: 232…233 °C
Point d'éclair: 250 °C (coupe fermée)
MDL Number: MFCD00013062
Numéro CAS: 54-64-8
ONU: 2025
ADR: 6.1,II
Merck Index: 12,09451
https://ch.vwr.com/stibo/web/std.lang.all/05/93/45390593.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR