Imprimer

Acide N-phénylanthranilique

ALEXALX550282G005EA 63 CHF
ALEXALX550282G005
Acide N-phénylanthranilique
Acide N-phénylanthranilique
Formule: C₁₃H₁₁NO₂
MW: 213,24 g/mol
Point d'ébullition: 375 °C (1013 hPa)
Melting Pt: 182…185 °C
MDL Number: MFCD00002421
Numéro CAS: 91-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR