Acide 2-acétoxybenzoïque
Synonyms:
Aspirin®
, Acide acétylsalicylíque
PROOMM0133.00EA
1830
CHF
PROOMM0133.00
Acide 2-acétoxybenzoïque
Acide 2-acétoxybenzoïque
Formule:
2-(CH₃CO₂)C₆H₄CO₂H MW: 180,16 g/mol Point d'ébullition: 140 °C (1013 hPa) Melting Pt: 138…140 °C Densité: 1,39 g/cm³ (20 °C) Point d'éclair: 250 °C |
MDL Number:
MFCD00002430 Numéro CAS: 50-78-2 Merck Index: 13,00856 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...