Imprimer

4-Chlororésorcinol

Fournisseur: Molekula

MOLEM11113179EA 0 CHF
MOLEM11113179
4-Chlororésorcinol
4-Chlororésorcinol
Formule: C₆H₅ClO₂
MW: 144,56 g/mol
Point d'ébullition: 259 °C (1013 hPa)
Melting Pt: 103…105 °C
MDL Number: MFCD00002273
Numéro CAS: 95-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR