Imprimer

2,4,6-Trichlorobenzonitrile

Fournisseur: Molekula

MOLE25180755-5GEA 0 CHF
MOLE25180755-5G
2,4,6-Trichlorobenzonitrile
2,4,6-Trichlorobenzonitrile
Formule: C₇H₂Cl₃N
MW: 206,46 g/mol
MDL Number: MFCD00052711
Numéro CAS: 6575-05-9
ONU: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR