Imprimer

2-Cyanobenzaldéhyde

Fournisseur: Molekula

MOLE25881995-5GEA 0 CHF
MOLE25881995-5G
2-Cyanobenzaldéhyde
2-Cyanobenzaldéhyde
Formule: C₈H₅NO
MW: 131,13 g/mol
MDL Number: MFCD00017503
Numéro CAS: 7468-67-9
ONU: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR