Imprimer

PAPA NONOate ≥97%

Fournisseur: Enzo Life Sciences
Attention

ALEXALX430016M005EA 0 CHF
ALEXALX430016M005 ALEXALX430016M025 ALEXALX4300165005
PAPA NONOate ≥97%
PAPA NONOate

Nitric oxide (NO) donor.


The half-life time of PAPA NONOate is 76.6 min at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Soluble in water or methanol. Relatively stable in alkaline solution.


Attention: Protect from moisture. Keep under inert gas.

Formule: C₆H₁₆N₄O₂
MW: 176,2 g/mol
température de stockage: Congélateur
Numéro CAS: 146672-58-4
https://ch.vwr.com/stibo/web/std.lang.all/06/42/45390642.jpg

Order Now

Résultats des tests de spécification

Purity ≥97%
Identity Determined by IR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR