GEA 5583 ≥97% (par HPLC)
Fournisseur: Enzo Life Sciences
ALEXALX430006M010EA
0
CHF
ALEXALX430006M010
ALEXALX430006M050
GEA 5583 ≥97% (par HPLC)
GEA 5583
Nitric oxide donor.
Stable nitric oxide releasing compound that is orally absorbed in rats.
Soluble in DMSO.
Résultats des tests de spécification
Purity | ≥97% (HPLC) |
Identity | Determined by IR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...