Imprimer

5-Nitroindoline-2,3-dione 97%

Attention

Synonyms: 5-Nitro-1H-indole-2,3-dione

ACRO128720250EA 104 CHF
ACRO128720250 ACRO128720050
5-Nitroindoline-2,3-dione 97%
5-Nitroindoline-2,3-dione
Formule: C₈H₄N₂O₄
MW: 192,13 g/mol
Melting Pt: 251 °C (déc.)
MDL Number: MFCD00005720
Numéro CAS: 611-09-6

Order Now

Résultats des tests de spécification

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥98.5 %
Stabilizer Approx. 500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR