Imprimer

DMNQ ≥99% (par CCM)

Fournisseur: Enzo Life Sciences
Attention

ALEXALX420027M025EA 0 CHF
ALEXALX420027M025 ALEXALX420027M005 ALEXALX420027M010
DMNQ ≥99% (par CCM)
DMNQ

Inducer of intracellular ROS generation.


Redox-cycling agent that induces intracellular superoxide anion formation and, depending on the concentration, induces cell proliferation, apoptosis or necrosis. DMNQ does not react with free thiol groups, is non-alkylating and adduct-forming in contrast to other quinones. Thus, DMNQ is a valuable tool for the generation of reactive oxygen species (ROS) in order to study the role of ROS in cell toxicity, apoptosis and necrosis.


Soluble in acetone, DMSO or methanol.

Formule: C₁₂H₁₀O₄
MW: 218,2 g/mol
température de stockage: Congélateur
MDL Number: MFCD00184331
Numéro CAS: 6956-96-3
https://ch.vwr.com/stibo/web/std.lang.all/06/16/45390616.jpg

Order Now

Résultats des tests de spécification

Purity ≥99% (TLC)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR