Imprimer
MGD sodium salt monohydrate ≥98% (par HPLC)
Fournisseur: Enzo Life Sciences
Attention
ALEXALX400014M050EA 0 CHF
ALEXALX400014M050 ALEXALX400014M250
MGD sodium salt monohydrate ≥98% (par HPLC)
MGD sodium salt monohydrate
Spin trapping reagent.
Together with FeSO₄ MGD is a useful component for the formation of the MGD2-Fe²⁺ complex, which is an excellent nitric oxide (NO) spin-trapping reagent. The MGD2-Fe²⁺ complex is quite unstable, especially in the presence of dissolved oxygen. Thus, the complex should be used immediately after being made. An excess (usually 5-fold excess), of MGD to Fe²⁺ is used for making the complex with FeSO₄ to give a more stable complex solution. Acidic conditions should be avoided because dithiocarbamate tends to decompose forming toxic carbon disulfide.
Soluble in water.
Formule: C₈H₁₆NO₅S₂Na·H₂O
MW: 311,3 g/mol
température de stockage: Réfrigérateur
Numéro CAS: 91840-27-6
https://ch.vwr.com/stibo/web/std.lang.all/05/94/45390594.jpg
Order Now
Résultats des tests de spécification
Purity ≥98% (HPLC)