Imprimer

DAR-4M 5 mM ≥98% (par HPLC)

Fournisseur: Enzo Life Sciences
Attention

ALEXALX620067M001EA 0 CHF
ALEXALX620067M001
DAR-4M 5 mM ≥98% (par HPLC)
DAR-4M

Fluorescent probe for NO detection.


Fluorescent probe (Ex: ~560 nm; Em: 575 nm) for the detection of nitric oxide (NO). It can be used to detect NO directly in biopsy tissues, cultured cells, and tissue sections. Stable in a pH range of 4 – 12. Low decrease in fluorescence intensity over time. Detection limit 7 nM.

Formule: C₂₅H₂₆N₄O₃
MW: 430,51 g/mol
température de stockage: Congélateur
Numéro CAS: 339527-79-6
EINECS: 200-664-3
https://ch.vwr.com/stibo/web/std.lang.all/08/79/45390879.jpg

Order Now

Résultats des tests de spécification

Concentration ~5 mM
Formulation Dissolved in 0.47 ml DMSO
Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR