Imprimer

Acide (2-bromophényl)acétique

Fournisseur: Molekula

MOLE10435778-25GEA 0 CHF
MOLE10435778-25G
Acide (2-bromophényl)acétique
Acide (2-bromophényl)acétique
Formule: (C₆H₄Br)CH₂COOH
MW: 215,05 g/mol
Point d'ébullition: 308 °C (1013 hPa)
Melting Pt: 107,5 °C
MDL Number: MFCD00004314
Numéro CAS: 18698-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR