Imprimer

Acide 2,2-diméthylglutarique ≥98%

Attention

B20481.14EA 160 CHF
B20481.14
Acide 2,2-diméthylglutarique ≥98%
Acide 2,2-diméthylglutarique
Formule: C₇H₁₂O₄
MW: 160,17 g/mol
Point d'ébullition: 180…200 °C (2 mmHg)
Melting Pt: 83…85 °C
MDL Number: MFCD00004197
Numéro CAS: 681-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR