Imprimer
ALEXALX746020
ALEXALX746020-T100EA 217 CHF
ALEXALX746020-T100
ALEXALX746020
TLR9 ligand.
CpG oligodeoxynucleotide (Type C) with phosphorothioate backbone. Specific ligand for human and mouse TLR9 (Toll-like receptor 9).
Sufficient for at least 100 cellular activation assays at 0.1 – 0.5 µg/well when performed in a 200 µl assay volume (i.e. 96 well microtiter plate).
Informations de livraison: Includes 1 vial of double distilled water (endotoxin-free).