Netropsin dihydrochloride ≥90% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Danger
|
ALEXALX380088M005EA
0
CHF
ALEXALX380088M005
Netropsin dihydrochloride ≥90% (par HPLC)
Netropsin dihydrochloride
Nucleic acid binding ligand.
Nucleic acid binding ligand. Binds to A-T-rich regions of the minor groove of DNA.
Soluble in alcohols or water.
Formule:
C₁₈H₂₆N₁₀O₃·2HCl MW: 503,5 g/mol température de stockage: Congélateur |
Numéro CAS:
1438-30-8 |
Résultats des tests de spécification
Purity | ≥90% (HPLC) |
Source | Isolated from Streptomyces netropsis |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...