Imprimer

1,2-Dioctanoyl-sn-glycerol (8:0) ≥96% (par CCM)

ALEXBML-DG112-0020EA 90 CHF
ALEXBML-DG112-0020 ALEXBML-DG112-0100
1,2-Dioctanoyl-sn-glycerol (8:0) ≥96% (par CCM)
1,2-Dioctanoyl-sn-glycerol (8:0)

PKC activator.


Potent activator of protein kinase C (PKC). Membrane permeable but non-physiologic DAG. Induces a discrete but transient translocation of protein kinase C.


Soluble in DMSO or 100% ethanol.

Formule: C₁₉H₃₆O₅
température de stockage: Congélateur ultra-basse température (–80°C)
Numéro CAS: 60514-48-9

Order Now

Résultats des tests de spécification

Purity ≥96% (TLC)
Identity Determined by ¹H-NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR