Imprimer

Zearalenone ≥99% (par HPLC, TLC)

Fournisseur: Enzo Life Sciences
Danger

ALEXALX630105M050EA 0 CHF
ALEXALX630105M050 ALEXALX630105M010
Zearalenone ≥99% (par HPLC, TLC)
Zearalenone

Mycotoxin.


Zearalenone (ZEN) is an estrogenic mycotoxin in animals and a phytohormone in plants. Inducer of sister chromatid exchange and chromosomal aberration. Acts as a protonophoric uncoupler in plant mitochondria.


Soluble in methanol (5 mg/ml).

Formule: C₁₈H₂₂O₅
MW: 318,4 g/mol
température de stockage: Congélateur
Numéro CAS: 17924-92-4
EINECS: 241-864-0
ONU: 2811
ADR: 6.1,II
https://ch.vwr.com/stibo/web/std.lang.all/08/95/45390895.jpg

Order Now

Résultats des tests de spécification

Source Isolated from Giberella zeae
Purity ≥99% (HPLC, TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR