Imprimer

Aflatoxin B1 ≥98% (par HPLC, TLC)

Fournisseur: Enzo Life Sciences
Danger

ALEXALX630093M001EA 0 CHF
ALEXALX630093M001 ALEXALX630093M005
Aflatoxin B1 ≥98% (par HPLC, TLC)
Aflatoxin B1

Naturally occurring mycotoxin produced by many species of Aspergillus. It is metabolised by the liver to the reactive intermediate aflatoxin M1. Exposure may produce acute necrosis, cirrhosis and carcinoma of the liver. Induces DNA damage.


Soluble in DMSO (20 mg/ml) or methylene chloride (10 mg/ml).

Formule: C₁₇H₁₂O₆
MW: 312,3 g/mol
température de stockage: Réfrigérateur
Numéro CAS: 1162-65-8
EINECS: 214-603-3
ONU: 2811
ADR: 6.1,I
https://ch.vwr.com/stibo/web/std.lang.all/08/89/45390889.jpg

Order Now

Résultats des tests de spécification

Source Isolated from Aspergillus flavus
Purity ≥98% (HPLC, TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR