Imprimer

Isoxanthohumol ≥98% (par 1H-RMN)

Fournisseur: Enzo Life Sciences
Attention

ALEXALX350279M001EA 0 CHF
ALEXALX350279M001
Isoxanthohumol ≥98% (par 1H-RMN)
Isoxanthohumol

Phytoestrogen.


Racemic mixture of (+/-)-Isoxanthohumol. Prenylated flavonoid. Induces apoptosis in mature adipocytes.


Soluble in DMSO (50 mg/ml) or methanol (50 mg/ml).

Formule: C₂₁H₂₂O₅
MW: 354,40 g/mol
température de stockage: Réfrigérateur
MDL Number: MFCD00238718
Numéro CAS: 521-48-2
ONU: 3077
ADR: 9,III
https://ch.vwr.com/stibo/web/std.lang.all/77/33/45347733.jpg

Order Now

Résultats des tests de spécification

Purity ≥98% (¹H-NMR)
Identity Determined by ¹H-NMR, ¹³C-NMR and LC-MS
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR