Imprimer

Kahweol ≥97% (par UPLC)

Fournisseur: Enzo Life Sciences

ALEXALX350223M010EA 0 CHF
ALEXALX350223M010
Kahweol ≥97% (par UPLC)
Kahweol

GST activator.


Induces glutathione S-transferase. Has anti-cancer, chemoprotective, anti-inflammatory and anti-atherosclerotic properties.


Soluble in ethyl acetate, acetone or DMSO; insoluble in water.

Formule: C₂₀H₂₆O₃
température de stockage: Congélateur
Numéro CAS: 6894-43-5
https://ch.vwr.com/stibo/web/std.lang.all/77/08/45347708.jpg

Order Now

Résultats des tests de spécification

Purity ≥97% (UPLC)
Source Natural product isolated from the unsaponifiable fraction of petroleum ether extract of coffee beans


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR