Imprimer
ALEXALX306033
ALEXALX306033-C500EA 308 CHF
ALEXALX306033-C500
ALEXALX306033
TCR activator.
Natural killer T cell (NKT) T cell receptor (TCR) agonist. Derivative of α-GalCer (the first defined and most potent agonistic antigen of TCR known so far). Stimulates the production of smaller amounts of IFN-γ than α-GalCer, but  induces equal or greater IL-4 secretion in mouse models.
Soluble in 100% ethanol, chloroform, DMSO, dimethyl formamide (warm), methanol or pyridine.