Imprimer

D-erythro-Sphingosine-1-phosphate (caged) ≥95%

Fournisseur: Enzo Life Sciences
Attention

ALEXALX306017C500EA 0 CHF
ALEXALX306017C500
D-erythro-Sphingosine-1-phosphate (caged) ≥95%
D-erythro-Sphingosine-1-phosphate (caged)

A photolysable derivative of sphingosine 1-phosphate (1-SPP). Important tool for the study of 1-SPP mediated intracellular events.


Hygroscopic and soluble in methanol or 100% ethanol.

https://ch.vwr.com/stibo/web/std.lang.all/76/29/45347629.jpg

Order Now

Résultats des tests de spécification

Purity ≥95%
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR