MK-571 sodium salt ≥90% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Attention
|
ALEXBML-RA109-0025EA
0
CHF
ALEXBML-RA109-0025
ALEXBML-RA109-0005
MK-571 sodium salt ≥90% (par HPLC)
MK-571 sodium salt
A potent, selective, orally active leukotriene D₄ antagonist. Ki=2.1 nM for inhibition of [³H]-LTD₄ binding to human lung membranes. Competitively antagonizes contractions of guinea pig trachea and ileum and human trachea induced by LTD₄. Blocks LTD₄ and ascaris-induced bronchoconstriction in conscious squirrel monkeys.
Soluble in water (20 mg/ml), aqueous buffers, 100% ethanol (10 mg/ml) or DMSO.
Formule:
C₂₆H₂₆ClN₂O₃S₂·Na MW: 537 g/mol température de stockage: Congélateur |
MDL Number:
MFCD11114392 Numéro CAS: 115103-85-0 |
Résultats des tests de spécification
Purity | ≥90% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...