AM 281 ≥98% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Danger
|
ALEXALX270240M005EA
0
CHF
ALEXALX270240M005
ALEXALX270240M025
ALEXALX270240M001
AM 281 ≥98% (par HPLC)
AM 281
CB1 receptor ligand.
Analog of the cannabinoid receptor (CB) antagonist SR 141716A. Potent CB1 receptor antagonist/inverse agonist (Ki = 14 nM).
Soluble in methanol (1 mg/ml) or DMSO (2 mg/ml at RT, 50 mg/ml in hot DMSO, after cooling this solution is stable over several days).
Formule:
C₂₁H₁₉Cl₂IN₄O₂ MW: 557,2 g/mol température de stockage: Réfrigérateur |
Numéro CAS:
202463-68-1 ONU: 2811 ADR: 6.1,III |
Résultats des tests de spécification
Purity | ≥98% (HPLC) |
Identity | Determined by ¹H-NMR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...