Imprimer
NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (par CCM)
Fournisseur: Enzo Life Sciences
Attention
ALEXALX106006M005EA 57 CHF
ALEXALX106006M005 ALEXALX106006M025
NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (par CCM)
NG, NG-Dimethyl-L-arginine dihydrochloride
NOS inhibitor.
Endogenous nitric oxide synthase (NOS) inhibitor.
Hygroscopic and soluble in water (50 mg/ml).
Formule: C₈H₁₈N₄O₂·2HCl·H₂O
température de stockage: Congélateur
Numéro CAS: 220805-22-1
https://ch.vwr.com/stibo/web/std.lang.all/74/25/45347425.jpg
Order Now
Résultats des tests de spécification
Purity ≥97% (TLC)