Imprimer
ALEXALX106005
ALEXALX106005M025EA 58 CHF
ALEXALX106005M025
ALEXALX106005
NOS inhibitor.
Inhibits nitric oxide synthesis. This salt form is more stable than the dihydrochloride.
Soluble in methanol.