Imprimer

Cholesterol-3-sulphate sodium salt

Fournisseur: Enzo Life Sciences

ALEXALX370009M005EA 0 CHF
ALEXALX370009M005 ALEXALX370009M025
Cholesterol-3-sulphate sodium salt
Cholesterol-3-sulphate sodium salt

PKC activator.


Activates multiple protein kinase C (PKC) iso-enzymes, especially the ε, η and ζ isoforms.


Soluble in methanol.

Formule: C₂₇H₄₅O₄S·Na
température de stockage: Réfrigérateur
Numéro CAS: 2864-50-8
https://ch.vwr.com/stibo/web/std.lang.all/03/95/45390395.jpg

Order Now

Résultats des tests de spécification

Purity One spot by TLC
Identity Determined by IR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR