N-Acetyl-leukotriene E4 ≥98% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Danger
|
ALEXBMLLEA004-0050EA
0
CHF
ALEXBMLLEA004-0050
N-Acetyl-leukotriene E4 ≥98% (par HPLC)
N-Acetyl-leukotriene E4
Major bilary metabolite of cysteinyl leukotrienes in the rat. Approximately 100-fold less active than LTC4. Equiactive with LTE4 in guinea pig lung parenchyma assay.
Soluble in ethanol.
Formule:
C₂₅H₃₉NO₆S température de stockage: Congélateur ultra-basse température (–80°C) |
Numéro CAS:
80115-95-3 ONU: 1992 ADR: 3,II |
Résultats des tests de spécification
Concentration | 50 µg/ml |
Purity | ≥98% (HPLC) |
Formulation | Solution in methanol/ammonium acetate buffer, 70:30. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...