D-myo-Inositol-1-monophosphate dipotassium salt ≥98% (par HPLC)
Fournisseur: Enzo Life Sciences
ALEXALX307026M005EA
0
CHF
ALEXALX307026M005
ALEXALX307026M001
D-myo-Inositol-1-monophosphate dipotassium salt ≥98% (par HPLC)
D-myo-Inositol-1-monophosphate dipotassium salt
Hygroscopic and soluble in water.
Formule:
C₆H₁₁O₉P·2K MW: 336,3 g/mol température de stockage: Congélateur |
Numéro CAS:
573-35-3 |
Résultats des tests de spécification
Purity | ≥98% by HPLC |
Identity | Determined by ¹H-NMR, ³¹P-NMR and MS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...