Imprimer

D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt ≥98% (par CCM)

Fournisseur: Enzo Life Sciences

ALEXALX307007M005EA 0 CHF
ALEXALX307007M005 ALEXALX307007M001
D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt ≥98% (par CCM)
D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt

Product of phosphatidylinositol phospholipid hydrolysis.


Causes release of calcium from intracellular stores.


Hygroscopic and soluble in water.

Formule: C₆H₁₂K₃O₁₅P₃
MW: 648,6 g/mol
température de stockage: Congélateur
Numéro CAS: 103476-24-0
https://ch.vwr.com/stibo/web/std.lang.all/76/34/45347634.jpg

Order Now

Résultats des tests de spécification

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR