OCH ≥95% (par CCM, RMN)
ALEXALX306029C250EA
224
CHF
ALEXALX306029C250
ALEXALX306029-M001
OCH ≥95% (par CCM, RMN)
OCH
Truncated analog of α-Gal-Cer.
Selectively induces IL-4, as opposed to IFN-γ. Offers protection in mouse against experimental autoimmune encephalomyelitis (EAE), an animal model of multiple sclerosis. Also offers protection against diabetes in NOD mice and against collagen induced arthritis. Stimulation of NKT cells with α-Gal-Cer results in release of a mixture of Th1 and Th2 cytokines, stimulation with OCH results in release of primarily Th2 cytokine.
Soluble in DMSO, 100% ethanol (warm) or methanol (warm).
Formule:
C₃₉H₇₇NO₉ MW: 704 g/mol température de stockage: Congélateur |
Numéro CAS:
383187-82-4 |
Résultats des tests de spécification
Purity | ≥95% (¹H-NMR and TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...