Leukotriene A4 methyl ester ≥95% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Danger
|
ALEXBML-LA004-0050EA
0
CHF
ALEXBML-LA004-0050
ALEXBML-LA004-1000
Leukotriene A4 methyl ester ≥95% (par HPLC)
Leukotriene A4 methyl ester
Substrate for LTA₄ hydrolase and LTC₄ synthase.
Unstable intermediate in the biosynthesis of LTB₄ and LTC₄. The naturally occuring free acid is a substrate for LTA₄ hydrolase and LTC₄ synthase and plays a central role in transcellular leukotrieneand lipoxin biosynthesis. Mobilises Ca²⁺ in human neutrophils. Supplied as the methyl ester for greater stability.
Formule:
C₂₁H₃₂O₃ température de stockage: Congélateur ultra-basse température (–80°C) |
Numéro CAS:
73466-12-3 ONU: 3077 ADR: 9,III |
Résultats des tests de spécification
Purity | ≥95% (HPLC) (remainder 11-trans isomer) |
Formulation | Oil dissolved in 2% triethylamine/hexane. |
Concentration | 1 mg/ml |
Source | Synthetic |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...