Imprimer

(R)-CPP ≥97% (par HPLC)

ALEXALX550231M001EA 43 CHF
ALEXALX550231M001
(R)-CPP ≥97% (par HPLC)
(R)-CPP

Highly potent NMDA receptor antagonist.


Soluble in dilute aqueous base or water.

Formule: C₈H₁₇N₂O₅P
température de stockage: Température ambiante
Numéro CAS: 126453-07-4

Order Now

Résultats des tests de spécification

Purity (HPLC) ≥97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR