Imprimer

FPL 64176 ≥98%

Fournisseur: Enzo Life Sciences

ALEXALX550229M005EA 0 CHF
ALEXALX550229M005
FPL 64176 ≥98%
FPL 64176

Ca²⁺ channel activator.


A potent L-type Ca²⁺ channel opener. Induces a contractile response in cardiac and vascular smooth muscle, EC₅₀=0.2 µM.


Hygroscopic and soluble in 100% ethanol (2 mg/ml) or DMSO (10 mg/ml); insoluble in aqueous solution.

Formule: C₂₂H₂₁NO₃
température de stockage: Réfrigérateur
Numéro CAS: 120934-96-5

Order Now

Résultats des tests de spécification

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR